Learn More
Biomatik Corporation Human Alpha-Crosslaps (aCTx) ELISA Kit
Supplier:  Biomatik Corporation EKU02283
Encompass_Preferred
Human Alpha-Crosslaps (aCTx) ELISA Kit for detecting aCTx in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 61.7-5000pg/ml. Sensitivity: 25.3pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Target Synonyms: a-CTXI; aCTX-I; A-Cr; Type I Collagen C-Telopeptide-Related Fraction. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4655