Learn More
Sino Biological Recombinant Human Alkaline Phosphatase/ALPL Protein (AVI Tag), Avi-tag Biotinylated, HPLC-verified
Catalog No. 502992910
Encompass_Preferred
A DNA sequence encoding the Human ALPL (NP_000469.3) (Met1-Ser502) was fused with an AVI tag at the C-terminus. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-299-2910 Supplier Sino Biological Supplier No. 10440H17HB100
A DNA sequence encoding the Human ALPL (NP_000469.3) (Met1-Ser502) was fused with an AVI tag at the C-terminus. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.