Learn More
Sino Biological Recombinant Human Alkaline Phosphatase/ALPL Protein (His Tag)
Supplier:  Sino Biological 10440H08H500
Encompass_Preferred
A DNA sequence encoding the human tissue-nonspecific alkaline phosphatase (NP_000469.3) (Met 1-Ser 502) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-255-5304