Learn More
Sino Biological Recombinant Human Alkaline Phosphatase/PLAP/ALPP Protein (His Tag), HPLC-verified
Catalog No. 502993124
Encompass_Preferred
A DNA sequence encoding the Human ALPP (P05187-1) (Ile23-Asp506) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-299-3124 Supplier Sino Biological Supplier No. 12387H08H1
A DNA sequence encoding the Human ALPP (P05187-1) (Ile23-Asp506) was expressed with a polyhistidine tag at the C-terminus.