Learn More
Sino Biological Recombinant Human Alkaline Phosphatase/PLAP/ALPP Protein (His Tag)
Supplier:  Sino Biological 12387H08Y500
Encompass_Preferred
A DNA sequence encoding human ALPP (AAH09647.1) (Ile23-Asp506) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-4374