Learn More
ACROBiosystems Human ALPP Protein, His Tag
Supplier:  ACROBiosystems ALPH52H3100UG
Encompass_Preferred
Human ALPP Protein, His Tag (ALP-H52H3) is expressed from human 293 cells (HEK293). It contains AA Ile 23 - Asp 506 (Accession # P05187-1).
Catalog No. 50-232-9112