Learn More
Sino Biological Anti-MIS/AMH Antibody, Mouse Monoclonal
Supplier:  Sino Biological 17500MM14100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human AMH (rh AMH; Catalog#17500-H07H; NP_000470.2; Leu25-Arg560). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-241-1349