Learn More
Sino Biological Recombinant Anti-MIS/AMH Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 17500R501100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human AMH (rh AMH; Catalog#17500-H07H; NP_000470.2; Leu25-Arg560).
Catalog No. 50-241-1282