Learn More
Sino Biological Anti-MIS/AMH Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 17500T24100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human AMH (rh AMH; Catalog#17500-H07H; NP_000470.2; Leu25-Arg560). AMH specific IgG was purified by Human AMH affinity chromatography.
Catalog No. 50-255-4006