Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human AMICA ELISA Kit

Catalog No. EH22RBX5
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH22RBX5 5 x 96 Tests
EH22RB 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EH22RBX5 Supplier Invitrogen™ Supplier No. EH22RBX5
Get Up to $3,000 in Extra Life Sciences Consumables with Purchase
View all promotions
Add to Cart
Add to Cart

ELISA

Human AMICA quantitates human AMICA in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human AMICA.

AMICA transmembrane protein of the plasma membrane of leukocytes that control their migration and activation through interaction with CXADR, a plasma membrane receptor found on adjacent epithelial and endothelial cells. The interaction between both receptors mediates the activation of gamma-delta T-cells, a subpopulation of T-cells residing in epithelia and involved in tissue homeostasis and repair. Upon epithelial CXADR-binding, JAML induces downstream cell signaling events in gamma-delta T-cells through PI3-kinase and MAP kinases. It results in proliferation and production of cytokines and growth factors by T-cells that in turn stimulate epithelial tissues repair. It also controls the transmigration of leukocytes within epithelial and endothelial tissues through adhesive interactions with epithelial and endothelial CXADR.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q86YT9
Assay Range 0.41-100 ng/mL
Assay Sensitivity 0.41 ng/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias adhesion molecule AMICA; adhesion molecule interacting with CXADR antigen 1; adhesion molecule, interacts with CXADR antigen 1; AMICA; Amica1; Crea7; CREA7-1; CREA7-4; Dendritic cell-specific protein CREA7; Dendritic cell-specific protein CREA7-1; dendritic-cell specific protein Crea7; dendritic-cell specific protein CREA7-1; dendritic-cell specific protein CREA7-4; Gm638; Jaml; junction adhesion molecule like; junctional adhesion molecule-like; mCrea7; UNQ722/PRO1387adhesion molecule AMICA; adhesion molecule interacting with CXADR antigen 1; adhesion molecule, interacts with CXADR antigen 1; AMICA; Amica1; Crea7; CREA7-1; CREA7-4; Dendritic cell-specific protein CREA7; Dendritic cell-specific protein CREA7-1; dendritic-cell specific protein Crea7; dendritic-cell specific protein CREA7-1; dendritic-cell specific protein CREA7-4; Gm638; Jaml; junction adhesion molecule like; junctional adhesion molecule-like; mCrea7; UNQ722/PRO1387;
Gene ID (Entrez) 120425
Gene Symbol JAML
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target AMICA
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.