Learn More
Sino Biological Recombinant Human AMIGO2 Protein (His Tag), Amine-Labeled Biotinylated
Supplier:  Sino Biological 13735H08HB20
Encompass_Preferred
A DNA sequence encoding the human AMIGO2 (NP_862830.1) (Met1-Thr398) was expressed with a polyhistidine tag at the C-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-255-4192