Learn More
Biomatik Corporation Human Amiloride Sensitive Sodium Channel Subunit Beta (SCNN1b) ELISA Kit
Supplier:  Biomatik Corporation EKU02306
Encompass_Preferred
Human Amiloride Sensitive Sodium Channel Subunit Beta (SCNN1b) ELISA Kit for detecting SCNN1b in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.107ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P51168. Gene ID: 6338. Target Synonyms: ENaCb; ENaCbeta; SCNEB; Liddle Syndrome; Sodium Channel,Nonvoltage-Gated 1,Beta; Epithelial Na (+) channel subunit beta; Nonvoltage-gated sodium channel 1 subunit beta. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7485