Learn More
Bioworld HUMAN AMINOACYLASE-1 (RECOM
Supplier:  Bioworld 220602441
Encompass_Preferred
HUMAN AMINOACYLASE-1 (RECOMBINANT): Recombinant Human AminoAcylase-1 (ACY1) Source: E.Coli Background : Aminoacylase-1 is a cytosolic, homodimeric, zinc-binding enzyme that catalyzes the hydrolysis of acylated L-amino acids to L-amino...
Catalog No. 50-253-344