Learn More
Biomatik Corporation Human Aminopeptidase B (RNPEP) ELISA Kit
Supplier:  Biomatik Corporation EKU12440
Encompass_Preferred
Human Aminopeptidase B (RNPEP) ELISA Kit for detecting RNPEP in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9H4A4. Gene ID: 6051. Target Synonyms: AP-B; APB; Aminopeptidase B; Arginine aminopeptidase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7449