Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Aminopeptidase LRAP/ERAP2 ELISA Kit

Catalog No. EH24RBX5
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH24RBX5 5 x 96 Tests
EH24RB 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EH24RBX5 Supplier Invitrogen™ Supplier No. EH24RBX5
Get Up to $3,000 in Extra Life Sciences Consumables with Purchase
View all promotions
Add to Cart
Add to Cart

ELISA

Human Aminopeptidase LRAP/ERAP2 quantitates human Aminopeptidase LRAP/ERAP2 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Aminopeptidase LRAP/ERAP2.

Aminopeptidases hydrolyze N-terminal amino acids of proteins or peptide substrates. Major histocompatibility complex class I molecules rely on aminopeptidases such as ERAP1 and LRAP to trim precursors to antigenic peptides in the endoplasmic reticulum following cleavage in the cytoplasm by tripeptidyl peptidase II.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q6P179
Assay Range 1.23-300 ng/mL
Assay Sensitivity 1.23 ng/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias Endoplasmic reticulum aminopeptidase 2; ERAP2; FLJ23633; FLJ23701; FLJ23807; Leukocyte-derived arginine aminopeptidase; LRAP; L-RAPEndoplasmic reticulum aminopeptidase 2; ERAP2; FLJ23633; FLJ23701; FLJ23807; Leukocyte-derived arginine aminopeptidase; LRAP; L-RAP;
Gene ID (Entrez) 64167
Gene Symbol ERAP2
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target ERAP2
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.