Learn More
Biomatik Corporation Human Amylase, Alpha 2A (AMY2A) ELISA Kit
Supplier:  Biomatik Corporation EKU09064
Encompass_Preferred
Human Amylase, Alpha 2A (AMY2A) ELISA Kit for detecting AMY2A in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40ng/ml. Sensitivity: 0.255ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P04746. Gene ID: 279. Target Synonyms: AMY2; AMY2B; PA; 1,4-alpha-D-glucan glucanohydrolase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8571