Learn More
Raybiotech Inc Human Amyloid beta 1-40 ELISA Kit
Supplier:  Raybiotech Inc ELHAMB1401
Encompass_Preferred
For Cell Culture Supernatants, Plasma, Serum, samples. Sensitivity: 0.1 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detects human Amyloid beta 1-40. It has crossreactivity with mouse Amyloid beta 1-40.
Catalog No. 50-194-8969