Learn More
TRIBIOSCIENCE INC Human Amyloid Beta 42 / Aβ42 ELISA Kit, 1 x 96-well plate
SDP
Catalog No. NC2886065
Encompass_Preferred
The Human Aβ42 ELISA is designed to quantitatively detect human Aβ42 levels in CSF, brain tissue, serum, plasma, and other biological samples. The kit uses proprietary methods to combine samples and detections in one step, rather than complex traditional methods. The detection range is from 78 to 5000 pg/mL.
Catalog No. NC2886065 Supplier TRIBIOSCIENCE INC Supplier No. TBS3299
The Human Aβ42 ELISA is designed to quantitatively detect human Aβ42 levels in CSF, brain tissue, serum, plasma, and other biological samples. The kit uses proprietary methods to combine samples and detections in one step, rather than complex traditional methods. The detection range is from 78 to 5000 pg/mL.