Learn More
Biomatik Corporation Human Amyloid Beta Peptide 1-42 (Ab1-42) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU12162
Encompass_Preferred
Human Amyloid Beta Peptide 1-42 (Ab1-42) Mini sample ELISA Kit for detecting Ab1-42 in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 3.70-300pg/ml. Sensitivity: 1.33pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P05067. Gene ID: 351. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8018