Learn More
Biomatik Corporation Human Amyloid Precursor Protein (APP) CLIA Kit
Supplier:  Biomatik Corporation EKU09923
Encompass_Preferred
Human Amyloid Precursor Protein (APP) CLIA Kit for detecting APP in serum, plasma, cerebrospinal fluid, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 6.86-5,000pg/ml. Sensitivity: 2.43pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P05067. Gene ID: 351. Target Synonyms: AAA; ABeta; ABPP; AD1; APPI; CVAP; PN2; CTFgamma; Peptidase Nexin-II Alzheimer Disease; Peptidase Nexin-II; Amyloid Beta (A4) Precursor Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7744