Learn More
IBL America Amyloid Beta (1-42) Aβ (Human) ELISA
Supplier:  IBL America 27711
Encompass_Preferred
Solid phase sandwich ELISA for the determination of human Aβ ( 1-42 ) in cerebrospinal fluid, cell culture media or the extract from brain tissue. For research use only, not for use in diagnostic procedures.
Catalog No. 50-212-0345