Learn More
Raybiotech Inc C-Series Human Angiogenesis Antibody Array 1 Kit.
Supplier:  Raybiotech Inc AAHANG12
Encompass_Preferred
Detects 20 Human Angiogenic Factors. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: Angiogenin, EGF, ENA-78 (CXCL5), bFGF, GRO alpha/beta/gamma, IFN-gamma, IGF-1, IL-6, IL-8 (CXCL8), Leptin, MCP-1 (CCL2), PDGF-BB, PLGF, RANTES (CCL5), TGF beta 1, TIMP-1, TIMP-2, Thrombopoietin (TPO), VEGF-A, VEGF-D
Catalog No. 50-194-8465