Learn More
Raybiotech Inc G-Series Human Angiogenesis Antibody Array 1000 Kit.
Catalog No. 501948472
Encompass_Preferred
A combination of Human Angiogenesis Antibody Array G1 & G2. Detects 43 Human Angiogenic Factors. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: Angiogenin, Angiopoietin-1, Angiopoietin-2, Angiostatin, bFGF, EGF, ENA-78 (CXCL5), Endostatin, GCSF, GM-CSF, GRO alpha/beta/gamma, I-309 (TCA-3/CCL1), IFN-gamma, IGF-1, IL-10, IL-1 alpha (IL-1 F1), IL-1 beta (IL-1 F2), IL-2, IL-4, IL-6, IL-8 (CXCL8), I-TAC (CXCL11), Leptin, MCP-1 (CCL2), MCP-3 (MARC/CCL7), MCP-4 (CCL13), MMP-1, MMP-9, PDGF-BB, PECAM-1 (CD31), PLGF, RANTES (CCL5), TGF beta 1, Thrombopoietin (TPO), Tie-2, TIMP-1, TIMP-2, TNF alpha, uPAR, VEGF-A, VEGFR2, VEGFR3, VEGF-D
Catalog No. 50-194-8472 Supplier Raybiotech Inc Supplier No. AAHANGG10008
A combination of Human Angiogenesis Antibody Array G1 & G2. Detects 43 Human Angiogenic Factors. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: Angiogenin, Angiopoietin-1, Angiopoietin-2, Angiostatin, bFGF, EGF, ENA-78 (CXCL5), Endostatin, GCSF, GM-CSF, GRO alpha/beta/gamma, I-309 (TCA-3/CCL1), IFN-gamma, IGF-1, IL-10, IL-1 alpha (IL-1 F1), IL-1 beta (IL-1 F2), IL-2, IL-4, IL-6, IL-8 (CXCL8), I-TAC (CXCL11), Leptin, MCP-1 (CCL2), MCP-3 (MARC/CCL7), MCP-4 (CCL13), MMP-1, MMP-9, PDGF-BB, PECAM-1 (CD31), PLGF, RANTES (CCL5), TGF beta 1, Thrombopoietin (TPO), Tie-2, TIMP-1, TIMP-2, TNF alpha, uPAR, VEGF-A, VEGFR2, VEGFR3, VEGF-D