Learn More
Raybiotech Inc G-Series Human Angiogenesis Array 1 Kit.
Catalog No. 501950358
Encompass_Preferred
Semi-quantitative measurement of 10 human cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: Angiogenin, Angiopoietin-2, EGF, bFGF, HB-EGF, HGF, Leptin, PDGF-BB, PLGF, VEGF-A
Catalog No. 50-195-0358 Supplier Raybiotech Inc Supplier No. GSHANG11
Semi-quantitative measurement of 10 human cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: Angiogenin, Angiopoietin-2, EGF, bFGF, HB-EGF, HGF, Leptin, PDGF-BB, PLGF, VEGF-A