Learn More
Raybiotech Inc G-Series Human Angiogenesis Array 2 Kit.
Supplier:  Raybiotech Inc GSHANG21
Encompass_Preferred
Semi-quantitative measurement of 30 human cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: Activin A, AgRP, Angiogenin, Angiopoietin-2, ANGPTL4, bFGF, ENA-78 (CXCL5), GRO alpha/beta/gamma, HB-EGF, HGF, IFN-gamma, IGF-1, IL-1 alpha (IL-1 F1), IL-2, IL-6, IL-8 (CXCL8), IL-17A, IP-10 (CXCL10), Leptin, LIF, MCP-1 (CCL2), PDGF-BB, PLGF, RANTES (CCL5), TGF beta 1, TIMP-1, TIMP-2, TNF alpha, TNF beta (TNFSF1B), Thrombopoietin (TPO)
Catalog No. 50-195-0360