Learn More
Raybiotech Inc G-Series Human Angiogenesis Array 3 Kit.
Supplier:  Raybiotech Inc GSHANG31
Encompass_Preferred
Semi-quantitative measurement of 30 human cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: Angiogenin, Angiostatin, CXCL16, EGF, FGF-4, Follistatin, GCSF, GM-CSF, I-309 (TCA-3/CCL1), IL-1 beta (IL-1 F2), IL-4, IL-10, IL-12 p40, IL-12 p70, I-TAC (CXCL11), MCP-2 (CCL8), MCP-3 (MARC/CCL7), MCP-4 (CCL13), MMP-1, MMP-9, PECAM-1 (CD31), TGF alpha, TGF beta 3, Tie-1, Tie-2, uPAR, VEGF-A, VEGFR2, VEGFR3, VEGF-D
Catalog No. 50-195-0362