Learn More
Raybiotech Inc Quantibody Human Angiogenesis Array 1 Kit.
Catalog No. 501950792
Encompass_Preferred
Detects 10 Human Angiogenic Factors. Suitable for all liquid sample types. Quantitative, Sandwich-based, Glass Slide Antibody Array. Simultaneous quantitative measurement of multiple proteins in a wide variety of sample types. Quantibody combines the high specificity and sensitivity of ELISA with the high throughput of the glass chip-based array. Less sample, more data: only 50 ul of sample is needed for quantification of up to 40 cytokines. Proteins detected: Angiogenin, Angiopoietin-2, EGF, bFGF, HB-EGF, HGF, Leptin, PDGF-BB, PLGF, VEGF-A
Catalog No. 50-195-0792 Supplier Raybiotech Inc Supplier No. QAHANG12
Detects 10 Human Angiogenic Factors. Suitable for all liquid sample types. Quantitative, Sandwich-based, Glass Slide Antibody Array. Simultaneous quantitative measurement of multiple proteins in a wide variety of sample types. Quantibody combines the high specificity and sensitivity of ELISA with the high throughput of the glass chip-based array. Less sample, more data: only 50 ul of sample is needed for quantification of up to 40 cytokines. Proteins detected: Angiogenin, Angiopoietin-2, EGF, bFGF, HB-EGF, HGF, Leptin, PDGF-BB, PLGF, VEGF-A