Learn More
Raybiotech Inc Quantibody Human Angiogenesis Array 2 Kit.
Catalog No. 501950794
Encompass_Preferred
Detects 30 Human Angiogenic Factors. Suitable for all liquid sample types. Quantitative, Sandwich-based, Glass Slide Antibody Array. Simultaneous quantitative measurement of multiple proteins in a wide variety of sample types. Quantibody combines the high specificity and sensitivity of ELISA with the high throughput of the glass chip-based array. Less sample, more data: only 50 ul of sample is needed for quantification of up to 40 cytokines. Proteins detected: Activin A, AgRP, Angiogenin, Angiopoietin-2, ANGPTL4, bFGF, ENA-78 (CXCL5), GRO alpha/beta/gamma, HB-EGF, HGF, IFN-gamma, IGF-1, IL-1 alpha (IL-1 F1), IL-2, IL-6, IL-8 (CXCL8), IL-17A, IP-10 (CXCL10), Leptin, LIF, MCP-1 (CCL2), PDGF-BB, PLGF, RANTES (CCL5), TGF beta 1, TIMP-1, TIMP-2, TNF alpha, TNF beta (TNFSF1B), Thrombopoietin (TPO)
Catalog No. 50-195-0794 Supplier Raybiotech Inc Supplier No. QAHANG22
Detects 30 Human Angiogenic Factors. Suitable for all liquid sample types. Quantitative, Sandwich-based, Glass Slide Antibody Array. Simultaneous quantitative measurement of multiple proteins in a wide variety of sample types. Quantibody combines the high specificity and sensitivity of ELISA with the high throughput of the glass chip-based array. Less sample, more data: only 50 ul of sample is needed for quantification of up to 40 cytokines. Proteins detected: Activin A, AgRP, Angiogenin, Angiopoietin-2, ANGPTL4, bFGF, ENA-78 (CXCL5), GRO alpha/beta/gamma, HB-EGF, HGF, IFN-gamma, IGF-1, IL-1 alpha (IL-1 F1), IL-2, IL-6, IL-8 (CXCL8), IL-17A, IP-10 (CXCL10), Leptin, LIF, MCP-1 (CCL2), PDGF-BB, PLGF, RANTES (CCL5), TGF beta 1, TIMP-1, TIMP-2, TNF alpha, TNF beta (TNFSF1B), Thrombopoietin (TPO)