Learn More
Raybiotech Inc Quantibody Human Angiogenesis Array 3 Kit.
Catalog No. 501950796
Encompass_Preferred
Detects 30 Human Angiogenic Factors. Suitable for all liquid sample types. Quantitative, Sandwich-based, Glass Slide Antibody Array. Simultaneous quantitative measurement of multiple proteins in a wide variety of sample types. Quantibody combines the high specificity and sensitivity of ELISA with the high throughput of the glass chip-based array. Less sample, more data: only 50 ul of sample is needed for quantification of up to 40 cytokines. Proteins detected: Angiogenin, Angiostatin, CXCL16, EGF, FGF-4, Follistatin, GCSF, GM-CSF, I-309 (TCA-3/CCL1), IL-1 beta (IL-1 F2), IL-4, IL-10, IL-12 p40, IL-12 p70, I-TAC (CXCL11), MCP-2 (CCL8), MCP-3 (MARC/CCL7), MCP-4 (CCL13), MMP-1, MMP-9, PECAM-1 (CD31), TGF alpha, TGF beta 3, Tie-1, Tie-2, uPAR, VEGF-A, VEGFR2, VEGFR3, VEGF-D
Catalog No. 50-195-0796 Supplier Raybiotech Inc Supplier No. QAHANG32
Detects 30 Human Angiogenic Factors. Suitable for all liquid sample types. Quantitative, Sandwich-based, Glass Slide Antibody Array. Simultaneous quantitative measurement of multiple proteins in a wide variety of sample types. Quantibody combines the high specificity and sensitivity of ELISA with the high throughput of the glass chip-based array. Less sample, more data: only 50 ul of sample is needed for quantification of up to 40 cytokines. Proteins detected: Angiogenin, Angiostatin, CXCL16, EGF, FGF-4, Follistatin, GCSF, GM-CSF, I-309 (TCA-3/CCL1), IL-1 beta (IL-1 F2), IL-4, IL-10, IL-12 p40, IL-12 p70, I-TAC (CXCL11), MCP-2 (CCL8), MCP-3 (MARC/CCL7), MCP-4 (CCL13), MMP-1, MMP-9, PECAM-1 (CD31), TGF alpha, TGF beta 3, Tie-1, Tie-2, uPAR, VEGF-A, VEGFR2, VEGFR3, VEGF-D