Learn More
Biomatik Corporation Human Angiopoietin 1 (ANGPT1) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU11854
Encompass_Preferred
Human Angiopoietin 1 (ANGPT1) Mini sample ELISA Kit for detecting ANGPT1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.24-15ng/ml. Sensitivity: 0.09ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q15389. Gene ID: 284. Target Synonyms: ANG1; AGP1; AGPT; ANG-I. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6102