Learn More
Biomatik Corporation Human Angiopoietin 2 (ANGPT2) CLIA Kit
Supplier:  Biomatik Corporation EKU10032
Encompass_Preferred
Human Angiopoietin 2 (ANGPT2) CLIA Kit for detecting ANGPT2 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 6.9-5,000pg/ml. Sensitivity: 2.9pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: O15123. Gene ID: 285. Target Synonyms: ANG2; AGPT2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8380