Learn More
ACROBiosystems ACROBiosystems Human Angiopoietin-like 3 / ANGPTL3 Protein, His Tag
Catalog No. 502106907
Encompass_Preferred
Recombinant Protein;1MG;Human Angiopoietin-like 3, His Tag (AN3-H52H3) is expressed from human 293 cells (HEK293). It contains AA Ser 17 - Glu 460 (Accession # Q9Y5C1-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 53.7 kDa.
Catalog No. 50-210-6907 Supplier ACROBiosystems Supplier No. AN3H52H31MG
Recombinant Protein;1MG;Human Angiopoietin-like 3, His Tag (AN3-H52H3) is expressed from human 293 cells (HEK293). It contains AA Ser 17 - Glu 460 (Accession # Q9Y5C1-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 53.7 kDa.