Learn More
American Research Products Inc Human angiopoietin like protei
Catalog No. 5000327245
Encompass
Human angiopoietin-like protein 8 (ANGPTL8) ELISA Kit from CUSABIO Detection Range 6 25 pg/mL-400 pg/mL Sensitivity 1 56 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27245 Supplier American Research Products Inc Supplier No. CSBEL028107HU
Human angiopoietin-like protein 8 (ANGPTL8) ELISA Kit from CUSABIO Detection Range 6 25 pg/mL-400 pg/mL Sensitivity 1 56 pg/mL Sample serum plasma tissue homogenates Method Sandwich