Learn More
Biomatik Corporation Human Angiotensin I (AngI) CLIA Kit
Supplier:  Biomatik Corporation EKU10028
Encompass_Preferred
Human Angiotensin I (AngI) CLIA Kit for detecting AngI in serum, plasma, tissue homogenates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 7.81-2,000pg/ml. Sensitivity: 2.92pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Chemiluminescence. Assay Time: 2h. Accession Number: P01019. Gene ID: 183. Target Synonyms: Ang-I; Angiotensin-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8381