Learn More
Biomatik Corporation Human Angiotensin II (AngII) CLIA Kit
Supplier:  Biomatik Corporation EKU10029
Encompass_Preferred
Human Angiotensin II (AngII) CLIA Kit for detecting AngII in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 11.72-3,000pg/ml. Sensitivity: 3.61pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Chemiluminescence. Assay Time: 2h. Accession Number: P01019. Gene ID: 183. Target Synonyms: Ang-II; Angiotensin-2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4839