Learn More
Raybiotech Inc Human Angiotensinogen (SERPINA8) ELISA Kit
Supplier:  Raybiotech Inc ELHAGT1
Encompass_Preferred
For Cell Culture Supernatants, Plasma, and Serum samples. Sensitivity: 1.22 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: The antibody pair provided in this kit recognizes Human Angiotensinogen (AGT/SERPINA8)
Catalog No. 50-194-8961