Learn More
Sino Biological Anti-Angiopoietin-1/ANGPT1 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 13667MM09100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Angiopoietin-1 (rh Angiopoietin-1; Catalog#13667-H02H1; NP_001137.2; Met1-Leu261 and Arg277-Phe498). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-255-4194