Learn More
Sino Biological Recombinant Human Angiopoietin-2/ANGPT2 Protein (mFc Tag)
Catalog No. 502992876
Encompass_Preferred
A DNA sequence encoding the Human ANGPT2 (NP_001138.1) (Tyr19-Phe496) was expressed with the Fc region of mouse IgG1 at the N-terminus.
Catalog No. 50-299-2876 Supplier Sino Biological Supplier No. 10691H04H100
A DNA sequence encoding the Human ANGPT2 (NP_001138.1) (Tyr19-Phe496) was expressed with the Fc region of mouse IgG1 at the N-terminus.