Learn More
Sino Biological Anti-Angiopoietin-2/ANGPT2 Antibody (HRP), Mouse Monoclonal
Supplier:  Sino Biological 10691MM21H100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Angiopoietin-2 (rh Angiopoietin-2; Catalog#10691-H08H; NP_001138.1; Met1-Phe496). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography and conjugated with horseradish-peroxidase (HRP).
Catalog No. 50-240-8304