Learn More
Sino Biological Human ANGPT2 MAb 100ul (Sino Biological)
Supplier:  Sino Biological 10691MM21100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Angiopoietin-2 (rh Angiopoietin-2; Catalog#10691-H08H; NP_001138.1; Met1-Phe496). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-3610