Learn More
Sino Biological Human ANGPT2 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 10691RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Angiopoietin-2 / ANG 2(rh Angiopoietin-2 / ANG 2; Catalog#10691-H08H; NP_001138.1; Met 1-Phe 496).Angiopoietin-2 / ANG 2 specific IgG was purified by Human Angiopoietin-2 / ANG 2 affinity chromatography.
Catalog No. 50-237-3386