Learn More
Sino Biological Human ANGPT2 Protein 100ug (Sino Biological)
Supplier:  Sino Biological 10691H27H1B100
Encompass_Preferred
A DNA sequence encoding the human Angiopoietin-2 (NP_001138.1) (Lys275-Phe496) was expressed with a c-terminal AVI tagged polyhistidine tag at the C-terminus. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-237-1395