Learn More
Sino Biological Recombinant Human Angiopoietin-2/ANGPT2 Protein (His Tag), Amine-Labeled Biotinylated
Supplier:  Sino Biological 10691H08SB100
Encompass_Preferred
A DNA sequence encoding the human ANGPT2 (NP_001138.1) (Met1-Phe496) was expressed with a polyhistidine tag at the C-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-255-5219