Learn More
Sino Biological Human ANGPT2 Protein 500ug (Sino Biological)
Supplier:  Sino Biological 10691H38H1500
Encompass_Preferred
A DNA sequence encoding the human ANGPT2 (NP_001138.1) (Met1-Gln248 and Lys275-Phe496 linked by a polypeptide linker) was expressed with the Fc region of mouse IgG2a at the C-terminus.
Catalog No. 50-237-2108