Learn More
Sino Biological Recombinant Human Angiopoietin-2/ANGPT2 Protein (mFc Tag)
Supplier:  Sino Biological 10691H38H150
Encompass_Preferred
A DNA sequence encoding the human ANGPT2 (NP_001138.1) (Met1-Gln248 and Lys275-Phe496 linked by a polypeptide linker) was expressed with the Fc region of mouse IgG2a at the C-terminus.
Catalog No. 50-255-5218