Learn More
Raybiotech Inc Human Angiopoietin-4 ELISA Kit
Supplier:  Raybiotech Inc ELHANGIOPOIETIN41
Encompass_Preferred
For Cell Culture Supernatants, Plasma, Serum, samples. Sensitivity: 0.25 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detects human Angiopoietin-4. Other species not determined.
Catalog No. 50-194-8976