Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human ANGPTL-3 Recombinant Protein, PeproTech®

Catalog No. 1301810UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
250 μg
50 μg
500 μg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
130-18-10UG 10 μg
130-18-1MG 1 mg
13018250UG 250 μg
13018500UG 500 μg
130-18-50UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 130-18-10UG Supplier Gibco™ Supplier No. 1301810UG
  • $92.50
    $69.65 / Each of 1

    Save $22.85

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Recombinant Protein

Recombinant human ANGPTL-3 is a glycoprotein that migrates by SDS-PAGE analysis at an apparent molecular weight of 62 kDa, and contains 452 amino acid residues including a C-terminal His tag. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

This gene encodes a member of a family of secreted proteins that function in angiogenesis. The encoded protein, which is expressed predominantly in the liver, is further processed into an N-terminal coiled-coil domain-containing chain and a C-terminal fibrinogen chain. The N-terminal chain is important for lipid metabolism, while the C-terminal chain may be involved in angiogenesis. Mutations in this gene cause familial hypobetalipoproteinemia type 2.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9Y5C1
For Use With (Application) Binding Assay, Bioactivity, Control, ELISA Standard
Formulation Protein with no preservative
Gene ID (Entrez) 27329
Molecular Weight (g/mol) 52.9 kDa
Name Human ANGPTL-3
Purification Method Purified
Quantity 10 μg
Source CHO cells
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias ANG-5; angiopoietin 5; angiopoietin like 3; Angiopoietin-5; angiopoietin-like 3; angiopoietin-like protein 3; Angiopoietin-related protein 3; ANGPT5; ANGPTL3; ANGPTL3(17-221); ANGPTL3(17-224); ANL3; FHBL2; hypl; hypolipidemia; UNQ153/PRO179
Common Name ANGPTL3
Gene Symbol ANGPTL3
Biological Activity Measured by its binding ability to recombinant alphavbeta3 integrin in a functional ELISA.
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Protein Tag His-tag
Sequence SRIDQDNSSF DSLSPEPKSR FAMLDDVKIL ANGLLQLGHG LKDFVHKTKG QINDIFQKLN IFDQSFYDLS LQTSEIKEEE KELRRTTYKL QVKNEEVKNM SLELNSKLES LLEEKILLQQ KVKYLEEQLT NLIQNQPETP EHPEVTSLKT FVEKQDNSIK DLLQTVEDQY KQLNQQHSQI KEIENQLRRT SIQEPTEISL SSKPRAPRTT PFLQLNEIRN VKHDGIPAEC TTIYNRGEHT SGMYAIRPSN SQVFHVYCDV ISGSPWTLIQ HRIDGSQNFN ETWENYKYGF GRLDGEFWLG LEKIYSIVKQ SNYVLRIELE DWKDNKHYIE YSFYLGNHET NYTLHLVAIT GNVPNAIPEN KDLVFSTWDH KAKGHFNCPE GYSGGWWWHD ECGENNLNGK YNKPRAKSKP ERRRGLSWKS QNGRLYSIKS TKMLIHPTDS ESFEHHHHHH HH
Content And Storage -20°C
Activity Measured by its binding ability to recombinant alphavbeta3 integrin in a functional ELISA.
Expression System CHO cells
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.