Learn More
Sino Biological Recombinant Human ANGPTL3/Angiopoietin-like 3 Protein (His Tag) 1 mg
Supplier:  Sino Biological 10770H08B1
Encompass_Preferred
A DNA sequence encoding the human ANGPTL3 (NP_055310.1) (Met1-Glu460) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-224-7066