Learn More
Raybiotech Inc Human ANGPTL3 ELISA Kit
Supplier:  Raybiotech Inc ELHANGPTL31
Encompass_Preferred
For serum, plasma, and cell culture supernatant samples. Sensitivity: 8.2 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA kit shows no cross-reactivity with the following cytokines tested: human beta IG-H3, CA9, Cathepsin B, CD23, CD26 (DPPIV), CHI3L1, Coagulation Factor III, CTLA-4, Dkk-4, EDA-A2, Erythropoietin R, FGF-6, FGF-9, FOLH1 (PSMA), Gas 1, IGFBP-5, IL-38 (IL-1 F10), IL-36 Ra (IL-1 F5), IL-36 alpha (IL-1 F6), IL-37 (IL-1 F7), IL-36 beta (IL-1 F8), IL-36 gamma (IL-1 F9), IL-1 R5 (IL-18 R alpha), IL-10 R alpha, IL-17C, IL-18, IL-20, IL-34, IL-5 R alpha, Layilin, Leptin R, Marapsin, Mer, MMP-7, P-Cadherin, Prostasin, SIGIRR, TGF beta R3,TWEAK.
Catalog No. 50-194-8978