Learn More
Sino Biological Recombinant Human ANGPTL3/Angiopoietin-like 3 Protein (His Tag)
Supplier:  Sino Biological 10770H08H21
Encompass_Preferred
A DNA sequence encoding the human ANGPTL3 (NP_055310.1) (Ser17-Pro220) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-5175